Skip to Content
MilliporeSigma
All Photos(2)

Documents

32543

Supelco

Oxfendazole-d3

VETRANAL®, analytical standard

Synonym(s):

(5-Benzenesulfinyl-1H-benzoimidazol-2-yl)carbamic acid methyl-d3 ester, Fenbendazole sulfoxide-d3, Methyl-d3 5-(phenylsulfinyl)benzimidazole-2-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D3H10N3O3S
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)c3ccccc3

InChI

1S/C15H13N3O3S/c1-21-15(19)18-14-16-12-8-7-11(9-13(12)17-14)22(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)/i1D3

InChI key

BEZZFPOZAYTVHN-FIBGUPNXSA-N

Application

Oxfendazole-d3 may be used as an internal standard for the determination of oxfendazole in bovine muscle tissue samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Femke Vandael et al.
Antibiotics (Basel, Switzerland), 9(9) (2020-09-04)
The vast majority of medicines in pig rearing are administered via oral group medication through medicated feed and drinking water. However, relevant on-farm factors affecting the concentration of these drugs in feed and drinking water, such as the homogeneity, stability
Multiresidue method for identification and quantification of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QuEChERS approach
Silva DRG, et al.
Talanta, 171, 307-320 (2017)
Xiao-Lin Hou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 962, 20-29 (2014-06-03)
A simple, sensitive and reliable analytical method was developed for the simultaneous determination of 38 veterinary drugs (18 sulfonamides, 11 quinolones and 9 benzimidazoles) and 8 metabolites of benzimidazoles in bovine milk by ultra high performance liquid chromatography-positive electrospray ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service