Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32487

Supelco

Spirotetramat Metabolite BYI08330 enol-glucoside

PESTANAL®, analytical standard

Synonym(s):

cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro [4.5]dec-3-en-4-yl β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33NO8
CAS Number:
Molecular Weight:
463.52
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

COC1CCC2(CC1)NC(=O)C(=C2O[C@@H]3O[C@H](CO)[C@H](O)[C@@H](O)[C@@H]3O)c4cc(C)ccc4C

InChI

1S/C24H33NO8/c1-12-4-5-13(2)15(10-12)17-21(24(25-22(17)30)8-6-14(31-3)7-9-24)33-23-20(29)19(28)18(27)16(11-26)32-23/h4-5,10,14,16,18-20,23,26-29H,6-9,11H2,1-3H3,(H,25,30)/t14?,16-,18-,19+,20-,23+,24?/m1/s1

InChI key

UZUGTDHNHPYPHX-WUMYBWKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Spirotetramat Metabolite BYI08330 enol-glucoside is one of the major metabolites of spirotetramat in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spirotetramat Metabolite BYI08330 enol-glucoside may be used as an analytical reference standard for the determination of spirotetramat metabolite BYI08330 enol-glucoside in:
  • Fruits and vegetables by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction based on liquid extraction/partition as well as dispersive solid phase extraction (d-SPE) and ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionization (ESI) source and multiple reaction monitoring (MRM) detection.
  • Pepper samples by chemometric-assisted QuEChERS extraction and UHPLC-ESI-MS/MS.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemometric-assisted QuEChERS extraction method for the residual analysis of thiacloprid, spirotetramat and spirotetramat?s four metabolites in pepper: application of their dissipation patterns.
Li S, et al.
Food Chemistry, 192, 893-899 (2016)
Simultaneous determination of spirotetramat and its four metabolites in fruits and vegetables using a modified quick, easy, cheap, effective, rugged, and safe method and liquid chromatography/tandem mass spectrometry.
Zhu Y, et al.
Journal of Chromatography A, 1299, 71-77 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service