Skip to Content
MilliporeSigma

32401

(±)-3-Chloro-1,2-propane-1,1,2,3,3-d5-diol

analytical standard

Synonym(s):

3-MCPD-d5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3D5H2ClO2
CAS Number:
Molecular Weight:
115.57
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])(O)C([2H])(O)C([2H])([2H])Cl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2/i1D2,2D2,3D

InChI key

SSZWWUDQMAHNAQ-UXXIZXEISA-N

General description

3-MCPD-d5 is the deuterated form of 3-MCDP.

Application

The internal standard can also be used in analyses as follows:
  • Determination of 3-monochloro-1,2-propanediol and 1,3-dichloro-2-propanol in various food product samples by liquid-liquid extraction (LLE) and gas chromatography-mass spectrometry (GC-MS)
  • Simultaneous analysis of 3-monochloropropanediol and glycidyl ester derivatives in fish oil samples using gas chromatography-tandem mass spectrometry (GC-MS/MS) following their derivatization with phenylboronic acid (PBA)
  • Development and validation of an ultra-high performance liquid chromatography-electrospray ionization-tandem triple quadrupole mass spectrometry (UHPLC-ESI-MS/MS) method for the measurement of 3-monochloropropane-1,2-diol in Caco-2 cells
  • Estimation of 3- and 2-monochloropropanediol esters in 10 extra virgin olive oil and 1 sunflower oil samples using an automated sample preparation procedure combined with GC-MS/MS
  • GC-MS method-based determination of total 3-MCPD concentration in edible fats and oils.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


signalword

Danger

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

target_organs

Kidney, Testes

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library