Skip to Content
MilliporeSigma

32124

Paraben internal standard mix solution

10 μg/mL in acetone, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

Paraben internal standard mix solution is a multi-standard mix solution for multi-analyte detection in the cosmetics industry since antioxidants and preservatives are often added in the cosmetics to extend the shelf life. These substances protect the products against damage and degradation to keep their unique aroma, color, or consistency.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Composition:
Methyl 4-hydroxybenzoate-ring-13C6
Ethyl 4-hydroxybenzoate-ring-13C6
Propyl 4-hydroxybenzoate-ring-13C6
Butyl 4-hydroxybenzoate-ring-13C6

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library