Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31716

Supelco

Thiocyclam hydrogenoxalate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NS3 · C2H2O4
CAS Number:
Molecular Weight:
271.38
Beilstein:
4771240
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)C(O)=O.CN(C)C1CSSSC1

InChI

1S/C5H11NS3.C2H2O4/c1-6(2)5-3-7-9-8-4-5;3-1(4)2(5)6/h5H,3-4H2,1-2H3;(H,3,4)(H,5,6)

InChI key

ICTQUFQQEYSGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiocyclam hydrogenoxalate is a derivative of nereistoxin, isolated from a marine annelid, Lubriconereis heteropoda Marenz. It is used as a pesticide for agricultural practices.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiocyclam hydrogenoxalate may be used as a reference standard in the determination of thiocyclam hydrogenoxalate in technical products and pharmaceutical formulations using reversed-phase high performance liquid chromatography with ultraviolet detection (RP-HPLC-UV).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>266.0 °F

Flash Point(C)

> 130 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Updated General Techniques and Additional Pesticides, 1150(1-2), 208-214 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service