Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31596

Supelco

Cycloxidim

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3S
CAS Number:
Molecular Weight:
325.47
Beilstein:
8436332
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CCC\C(=N/OCC)C1=C(O)CC(CC1=O)C2CCCSC2

InChI

1S/C17H27NO3S/c1-3-6-14(18-21-4-2)17-15(19)9-13(10-16(17)20)12-7-5-8-22-11-12/h12-13,19H,3-11H2,1-2H3/b18-14+

InChI key

GGWHBJGBERXSLL-NBVRZTHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cycloxidim belongs to cyclohexanedione group and is an active selective ingredient used in herbicides.

Application

Cycloxidim has been used as working standard in the determination residual pesticides in rice using liquid chromatography-tandem mass spectrometry with electrospray ionization in positive mode (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of twenty pesticides in rice by employing QuEChERS and LC-ESI-MS/MS.
Rebelo, Andrey M., et al.
Analytical Methods : Advancing Methods and Applications, 6.23, 9469-9476 (2014)
The effect of cycloxydim tolerant maize (CTM) alleles on grain yield and agronomic traits of maize single cross hybrid.
Vancetovic, J., et al.
Maydica, 54.1 , 91-91 (2009)
Shirin Monadjemi et al.
Chemosphere, 89(3), 269-273 (2012-05-19)
The reactivity of the herbicide cycloxydim (CD) toward singlet oxygen was studied in organic solution and on wax films to mimic the leaf surface. Experiments in solution were conducted in acetonitrile using phenalenone as a sensitizer. For the experiments in
Cafer Turgut
Environmental science and pollution research international, 12(6), 342-346 (2005-11-25)
Aquatic plants have a great potential to function as in situ, on-site biosinks and biofilters of pollutants. They are used for phytoremediation and phytotoxicity studies. Pesticide uptake studies are very important to predict contaminant accumulation, translocation, and transformation. There are
K M Cocker et al.
Pest management science, 57(7), 587-597 (2001-07-24)
Herbicide-resistant Lolium multiflorum (Italian rye-grass) was first reported in the UK in 1993 and had been confirmed on 25 farms by 1999. In this study, resistance to five herbicides belonging to the aryloxyphenoxypropionate, cyclohexanedione and phenyl-urea classes was determined in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service