Skip to Content
MilliporeSigma
All Photos(1)

Documents

31306

Supelco

Benzo[a]pyrene solution

100 μg/mL in cyclohexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)cc3ccc4cccc5ccc2c3c45

InChI

1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H

InChI key

FMMWHPNWAFZXNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo[a]pyrene belongs to the family of polycyclic aromatic hydrocarbon (PAH) compounds, which consist of two or more aromatic rings made up of only hydrogen and carbon elements.

Application

Benzo[a]pyrene may be used as a reference standard for the determination of the analyte in olive oil samples by gas chromatography-mass spectrometry (GC-MS) as well as liquid chromatography-fluorescence detection techniques (LC-FLD), and in cereal-based foodstuffs, fish, meat and poultry by GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High-throughput analytical method for 265 pesticides and environmental contaminants in meats and poultry by fast low pressure gas chromatography and ultrahigh-performance liquid chromatography tandem mass spectrometry
Sapozhnikova Y
Journal of Chromatography A, 1572(1-2), 203-211 (2018)
Multi-class, multi-residue analysis of pesticides, polychlorinated biphenyls, polycyclic aromatic hydrocarbons, polybrominated diphenyl ethers and novel flame retardants in fish using fast, low-pressure gas chromatography-tandem mass spectrometry
Sapozhnikova Y and Lehotay SJ
Analytica Chimica Acta, 758(1-2), 80-92 (2013)
Assessing polycyclic aromatic hydrocarbons in cereal-based foodstuffs by using a continuous solid-phase extraction system and gas chromatography-mass spectrometry
Rascon AJ, et al.
Food Control, 92(1-2), 92-100 (2018)
Direct analysis of benzo[a]pyrene metabolites with strong overlapping in both the spectral and lifetime domains
Alfarhani B, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 137, 51-61 (2018)
Rapid determination of benzo(a)pyrene in olive oil samples with solid-phase extraction and low-pressure, wide-bore gas chromatography-mass spectrometry and fast liquid chromatography with fluorescence detection
Bogusz MJ, et al.
Journal of Chromatography A, 1026(1-2), 1-7 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service