Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30673

Supelco

Thiacloprid-(thiazolidin ring-d4)

PESTANAL®, analytical standard

Synonym(s):

Thiacloprid-d4, [3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidin-d4-ylidene]cyanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D4H5ClN4S
Molecular Weight:
256.75
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

ClC1=CC=C(CN2C([2H])([2H])C([2H])([2H])S/C2=N\C#N)C=N1

InChI

1S/C10H9ClN4S/c11-9-2-1-8(5-13-9)6-15-3-4-16-10(15)14-7-12/h1-2,5H,3-4,6H2/b14-10-/i3D2,4D2

InChI key

HOKKPVIRMVDYPB-NPNQNPJSSA-N

General description

Find more information here: Neonicotinoids

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service