Skip to Content
MilliporeSigma

29680

Cyclopentane

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H10
CAS Number:
Molecular Weight:
70.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
206-016-6
Beilstein/REAXYS Number:
1900195
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

~2 (vs air)

assay

≥99.0% (GC)

autoignition temp.

682 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.7 %

technique(s)

HPLC: suitable, chromatography: suitable, gas chromatography (GC): suitable

impurities

cyclopentene

refractive index

n20/D 1.405 (lit.), n20/D 1.406

bp

50 °C (lit.)

mp

−94 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

General description

Cyclopentane is a colourless flammable acyclic hydrocarbon liquid. It is a widely used component in preparing products like analgesics, insecticides, sedatives, antitumor agents and also finds application in pharmaceutical industry.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?

Explore all of our products under Cyclopentane


Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

-4.0 °F - closed cup

Flash Point (°C)

-20 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library