Skip to Content
MilliporeSigma

29231

Chloramphenicol Selective Supplement

powder, for yeasts, for molds, pack of 5 vials

Synonym(s):

Antibiotic supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171614
EC Number:
200-287-4
NACRES:
NA.85
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pack of 5 vials

application(s)

environmental
food and beverages
microbiology
pharmaceutical

storage temp.

2-8°C

suitability

molds, yeasts

General description

Chloramphenicol Selective Supplement is a sterile, powdered antibiotic additive containing 50 mg of chloramphenicol per vial. It is specifically designed for microbiological applications to inhibit bacterial growth while allowing the selective isolation of yeasts and molds. Each vial is sufficient for preparing 500 mL of medium, making it ideal for use in environmental, food, beverage, and pharmaceutical testing.

Application

Chloramphenicol Selective Supplement is widely used for the selective isolation of yeasts and molds by suppressing bacterial growth, enabling the detection and enumeration of fungal species in mixed cultures. It is applied in microbiological testing across food safety, environmental monitoring, and pharmaceutical quality control to ensure accurate identification of fungal contaminants.

Packaging

package with 5 vials

Other Notes

(A vial is sufficient for 500 mL of medium)
Chloramphenicol 50 mg


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library