Skip to Content
MilliporeSigma
All Photos(1)

Documents

234079

Sigma-Aldrich

Erythrosin yellowish blend

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

powder or crystals

Quality Level

composition

Eosin Y, 10%
Erythrosin B, 90%

solubility

H2O: 10 mg/mL, clear, red

λmax

525 nm

Amax

≥0.12 at 303-309 nm
≥0.25 at 255-261 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service