Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21049

Supelco

Calcium standard for AAS

analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM

Synonym(s):

Calcium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

1.000 g/L Ca+2 in hydrochloric acid

technique(s)

AAS: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

SMILES string

Cl[Ca]Cl

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium standard for AAS is used as an analytical reference standard for the detection of the analyte in green/red cabbage samples using flame photometric analysis.

Preparation Note

prepared with CaCl2.2H2O and HCl (~0.5 M)

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compositional analysis of Pakistani green and red cabbage
Ashfaq F, et al.
Pakistan Journal of Agricultural Sciences, 55(1), The-The (2018)
Wei-Chuan Chen et al.
Bioresource technology, 291, 121891-121891 (2019-08-07)
This work studies a series of strategies in the production of lutein by Scenedesmus obliquus CWL-1 under mixotrophic cultivation. Our experimental results revealed that the optimal conditions associated with light-related strategies were 12 h light period followed by a 12 h dark
Daniela Predoi et al.
Polymers, 11(5) (2019-05-18)
Dextran-coated zinc-doped hydroxyapatite (ZnHApD) was synthesized by an adapted sol-gel method. The stability of ZnHApD nanoparticles in an aqueous solution was analyzed using ultrasonic measurements. The analysis of the evolution in time of the attenuation for each of the frequencies
Xiaoyu Du et al.
Materials science & engineering. C, Materials for biological applications, 103, 109731-109731 (2019-07-28)
The fabrication of bone tissue engineering scaffolds with high osteogenic ability and favorable mechanical properties is of huge interest. In this study, a silk fibroin (SF) solution of 30 wt% was extracted from cocoons and combined with mesoporous bioactive glass (MBG)
Marie Dubus et al.
Colloids and surfaces. B, Biointerfaces, 181, 671-679 (2019-06-22)
The regeneration of bone-soft tissue interface, using functional membranes, remains challenging and can be promoted by improving mesenchymal stem cells (MSCs) paracrine function. Herein, a collagen membrane, used as guided bone regeneration membrane, was functionalized by calcium phosphate, chitosan and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service