Key Documents
20683
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1025TLL, Kel-F CTFE Luer lock, volume 25 mL, needle size 22 gauge (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
material
Kel-F CTFE Luer lock
needle size
22 gauge (not included)
feature
syringe type TLL (Luer Lock) without slots (needle not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 82520
needle L
, n/a
volume
25 mL
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192-192 (2012)