Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205435

Sigma-Aldrich

May-Grünwald Stain

Suitable for the differential staining of cellular elements of blood

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Quality Level

form

powder

technique(s)

microbe id | staining: suitable

color

green to black

λmax

522 nm (2nd)
645 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COC1=NC(=NC2=C1N=CN2C3C(C(C(O3)CO)O)O)N

InChI

InChI=1S/C11H15N5O5/c1-20-9-5-8(14-11(12)15-9)16(3-13-5)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2,12,14,15)/t4-,6+,7?,10-/m1/s1

InChI key

IXOXBSCIXZEQEQ-PKJMTWSGSA-N

General description

May-Grünwald stain is a Romanowsky neutraldye stain based on the combination ofmethylene blue azures as the basic dyecomponent and eosin as the acid dyecomponent.It is intended for use in staining blood films orbone marrow films. It is used routinely for diagnostic purposes in hematology.

Application

May-Grünwald′s stain is used for the differential staining of cellular elements of blood.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preclinical evaluation of engineered oncolytic herpes simplex virus for the treatment of pediatric solid tumors
Megison ML, et al.
PLoS ONE, 9(1), e86843-e86843 (2014)
Michael L Megison et al.
PloS one, 9(1), e86843-e86843 (2014-02-06)
Recently, investigators showed that mice with syngeneic murine gliomas that were treated with a neuroattenuated oncolytic herpes simplex virus-1 (oHSV), M002, had a significant increase in survival. M002 has deletions in both copies of the γ134.5 gene, enabling replication in
Arumugam Ponnan et al.
Environmental science and pollution research international, 27(25), 31760-31766 (2020-06-07)
Fucoidan is a unique bioactive and dietary polymer enriched mainly in the cell wall matrix of the brown seaweeds. This present study was intended to reveal the antigenotoxicity effect of fucoidan on 4-nitroquinolin-1-oxide (4-NQO) induced genetics damage and apoptosis in
Epidemiological characteristics of Malassezia folliculitis and use of the May-Grunwald-Giemsa stain to diagnose the infection
Durdu, et al.
Diagnostic Microbiology and Infectious Disease, 76(4) (2013)
Historical hematology: May--Hegglin anomaly
Saito, Hidehiko and Kunishima, Shinji
American Journal of Hematology, 83(4) (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service