Skip to Content
MilliporeSigma

19945

Oleoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(9Z)-1-oxo-9-octadecen-1-yl]oxy]-1-propanaminium inner salt, cis-9-Octadecenoyl-L-carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H47NO4
CAS Number:
Molecular Weight:
425.64
UNSPSC Code:
41116107
NACRES:
NB.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

≥95.0% (HPLC)

optical activity

[α]/D -13.0±2.0°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCC/C=C\CCCCCCCC)=O)CC([O-])=O

InChI

1S/C25H47NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h12-13,23H,5-11,14-22H2,1-4H3/b13-12-/t23-/m1/s1

InChI key

IPOLTUVFXFHAHI-WHIOSMTNSA-N

General description

Oleoyl-L-carnitine belongs to the class of long-chain acylcarnitines, the fatty acyl esters of L-carnitine. Its role as a potential analgesic agent and ability to inhibit the glycine transport by GlyT2 transporter is investigated.

Application

Oleoyl-L-carnitine is used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled to mass spectrometry (LC-MS) technique.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library