Skip to Content
MilliporeSigma

18645

Antonia Red Dextran 40

Mw 35000-55000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12171501
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Leuconostoc mesenteroides B-512F)

form

powder or crystals

mol wt

Mw 35000-55000

manufacturer/tradename

TdB Consultancy AB

extent of labeling

0.001-0.01 (degree of substitution)

fluorescence

λex 583 nm; λem 602 nm in borate buffer (pH 9.0)

detection method

fluorometric

storage temp.

room temp

General description

Antonia Red dextrans are purple powders that are readily soluble in water or electrolytesolutions. Antonia Red dextran are insoluble in most organic solvents.

Application

Antonia Red dextranes are primarily used for studying permeability and transport in cells and tissues, trace cell lineage and perform vasculature studies. An added benefit is that measurements of the fluorescence provide quantitative data on the permeability of healthy and diseased tissues. Such studies can be performed in real time by intra-vital fluorescence microscopy. The technique offers high sensitivity and concentrations down to 1μg/ml can be detected in tissue fluids. Antonia Red dextranes is brightly fluorescent using the red (Texas Red®) filter in fluorescence microscopy.

Other Notes

The dextran used is essentially a linear α-(1-6)-linked glucose chain with a low percentage (2-5%) of α-(1-3) branches distributed along the chain.

Legal Information

Antonia Red is a trademark of TdB Consultancy AB
Texas Red is a registered trademark of Life Technologies


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library