Skip to Content
MilliporeSigma

18477

N-(2,6-Dichlorophenyl)-2-indolinone

analytical standard

Synonym(s):

1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, NSC 621845

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H9Cl2NO
CAS Number:
Molecular Weight:
278.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
239-399-3
Beilstein/REAXYS Number:
1538309
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

API family

diclofenac

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(N2C(CC3=C2C=CC=C3)=O)C(Cl)=CC=C1

InChI

1S/C14H9Cl2NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(17)18/h1-7H,8H2

InChI key

JCICIFOYVSPMHG-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library