Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18143

Supelco

trans-Nerolidol

analytical standard

Synonym(s):

(E)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, trans-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
Beilstein:
5731231
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.479 (lit.)

bp

145-146 °C/12 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

C\C(C)=C/CC\C(C)=C\CCC(C)(O)C=C

InChI

1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+

InChI key

FQTLCLSUCSAZDY-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nerolidol is a naturally occurring sesquiterpene found in the essential oils.

Application

It has been used as reference sample for analyzing sterols and related compounds in root bark of Oplopanax horridus using HPLC and TLC methods.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatography and thin-layer chromatography assays for Devil's Club (Oplopanax horridus).
Gruber JW
Journal of Chromatographic Science, 42(4), 196-199 (2004)
Fernanda Pículo et al.
Journal of applied toxicology : JAT, 31(7), 633-639 (2010-11-23)
Nerolidol is a sesquiterpenoid component of essential oil used as a flavor and aroma enhancer. It has also been studied as a topical skin penetration enhancer, and has inhibitory activities against S. aureus and E. coli, among other activities. The
Shigeru Tamogami et al.
FEBS letters, 585(12), 1807-1813 (2011-04-23)
DMNT biosynthesis was proposed to proceed via (E)-nerolidol in plants a decade ago. However, (E)-nerolidol function as airborne signal/substrate for in-vivo biosynthesis of DMNT remains to be investigated and the regulation of DMNT production and emission is largely unknown. We
Mahmoud AbouLaila et al.
Parasitology international, 59(2), 278-282 (2010-02-25)
Nerolidol is a sesquiterpene present in the essential oils of many plants, approved by the U.S. FDA as a food flavoring agent. Nerolidol interferes with the isoprenoid biosynthetic pathway in the apicoplast of P. falciparum. In the present study, the
Fernanda M Ferreira et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(2), 189-196 (2011-12-06)
In the present work, we evaluated the potential toxic effects of nerolidol, a sesquiterpenoid common in plants essential oils, both on mitochondrial and cellular energetics. Samples of enriched natural extracts of nerolidol (a racemic mixture of cis and trans isomers)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service