Skip to Content
MilliporeSigma
All Photos(2)

Documents

17173

Millipore

Rappaport Vassiliadis Broth, modified

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Magnesium chloride Malachite Green Broth, Modified Rappaport Vassiliadis Broth, RV Broth, Salmonella Enrichment Broth acc. to Rappaport and Vassiliadis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to ISO 19250:2010
according to ISO 6579-1:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.2±0.2 (25 °C)

application(s)

environmental
food and beverages
food and beverages
pathogen testing

microbiology

suitability

selective for Salmonella spp.

Application

Used as a selective enrichment medium for the isolation of Salmonella species from food and environmental specimens.

Components

Papaic digest of soyabean meal 5 g/1110 mL;
Sodium chloride 8 g/1110 mL;
Monopotassium phosphate 1.6 g/1110 mL;
Magnesium chloride anhydrous 18.7 g/l (is equivalent to 40 g/1110 mL Magnesium chloride hexahydrate);
Malachite Green 0.04 g/1110 ml

Preparation Note

Suspend 30.0 g in 1 litre distilled water. Heat gently if necessary to dissolve the medium completely. Dispense as desired into tubes and sterilize by autoclaving at 115°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service