Skip to Content
MilliporeSigma
All Photos(1)

Documents

17137

Millipore

CPC Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile

Quality Level

form

powder

application(s)

food and beverages
food and beverages

microbiology

storage temp.

2-8°C

suitability

Vibrio spp.

Application

For selective isolation and enumeration of vibrio species from foods.

Components

Composition (per vial; sufficient for 500 mL medium)
Colistin methane sulfonate 63 mg; Polymyxin B sulfate 9 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service