Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16654

Supelco

Quercetin 3-glucoside

analytical standard

Synonym(s):

Quercetin 3-β-D-glucoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
100989
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-QSOFNFLRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin is a flavonoid that can be consumed in the diet. It has antioxidant, anti-inflammatory, antiproliferative and anticarcinogenic properties. Quercetin 3-glucoside (Q3G) is a glucoside derivative of quercetin.[1]

Application

Q3G may be used as standard for calibration during HPLC analysis of diverse grape and wine phenolics using direct injection with UV-Vis photodiode array (DAD) and fluorescence detection.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: allium angelica arnica carum eucalyptus ginkgo hedera humulus hypericum melissa pimpinella primula sambucus glycyrrhiza lycium serenoa serenoa
This compound is commonly found in plants of the genus: arnica sambucus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC analysis of diverse grape and wine phenolics using direct injection and multidetection by DAD and fluorescence.
Gomez-Alonso, Sergio, Esteban Garcia-Romero, and Isidro Hermosin-Gutierrez.
J. Food Compos. Anal., 20.7, 618-626 (2007)
Aloys L A Sesink et al.
The Journal of nutrition, 133(3), 773-776 (2003-03-04)
Quercetin has antioxidant, anti-inflammatory, antiproliferative and anticarcinogenic properties. In plant foods, quercetin occurs mainly bound to various sugars via a beta-glycosidic link. We hypothesized that lactase phlorizin hydrolase (LPH), an enzyme at the brush border membrane of intestinal cells, is
Determination of the metabolite content of Brassica juncea cultivars using comprehensive two-dimensional liquid chromatography coupled with a photodiode array and mass spectrometry detection
Arena K, et al.
Molecules (Basel), 25(5), 1235-1235 (2020)
Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
Zhongxiang Zhao et al.
Journal of natural products, 70(10), 1683-1686 (2007-10-10)
Five new flavan-4-ol glycosides, abacopterins E-I ( 5- 9), and seven known flavonoid glycosides ( 3 and 10- 15) were isolated from the aerial parts of the fern Abacopteris penangiana. Their structures were elucidated on the basis of extensive spectroscopic

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service