Skip to Content
MilliporeSigma

14765

Thallium(I) acetate

≥97.0%, solid

Synonym(s):

TlOAc, Acetic acid thallium(I) salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3COOTl
CAS Number:
Molecular Weight:
263.43
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12161600
EC Number:
209-257-5
MDL number:
Beilstein/REAXYS Number:
3693914
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Thallium(I) acetate, purum, ≥97.0%

grade

purum

Quality Segment

assay

≥97.0%

form

solid

reaction suitability

core: thallium, reagent type: catalyst

SMILES string

CC(=O)O[Tl]

InChI

1S/C2H4O2.Tl/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

HQOJMTATBXYHNR-UHFFFAOYSA-M

General description

Thallium (I) acetate functions as a catalyst to enhance the rate of chemical reactions in the synthesis of isothiocyanates from isocyanides and disulfides. In addition, it is also used as the source of thallium ions in organic solvents for electrochemical reactions.

Application

  • Thallium Use: An Overview: Offers a comprehensive overview of Thallium(I) acetate′s use in organic synthesis in the oxidation of olefins and hydrocarbons, and in epoxidation and polymerization reactions (Genchi et al., 2021).


Still not finding the right product?

Explore all of our products under Thallium(I) acetate


Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library