Skip to Content
MilliporeSigma

13259

Dimethyl carbonate

greener alternative

analytical standard

Synonym(s):

Carbonic acid dimethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
UNSPSC Code:
77101502
NACRES:
NA.24
PubChem Substance ID:
EC Number:
210-478-4
Beilstein/REAXYS Number:
635821
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

3.1 (vs air)

vapor pressure

18 mmHg ( 21.1 °C)

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤0.02% water

refractive index

n20/D 1.368 (lit.), n20/D 1.369

bp

90 °C (lit.)

mp

2-4 °C (lit.)

density

1.069 g/mL at 25 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

greener alternative category

SMILES string

O=C(OC)OC

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3

InChI key

IEJIGPNLZYLLBP-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a Green chemical. Click here for more information.

Dimethyl carbonate is a unique molecule with versatile chemical reactivity. It is an environmentally benign building block and also a safe substitute for dimethyl sulfate or methyl halides.


Still not finding the right product?

Explore all of our products under Dimethyl carbonate


pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library