Skip to Content
MilliporeSigma

09985

Ammonium tartrate dibasic

BioUltra, ≥98% (T)

Synonym(s):

L-(+)-Tartaric acid diammonium salt, Diammonium tartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2C4H4O6
CAS Number:
Molecular Weight:
184.15
NACRES:
NA.25
PubChem Substance ID:
eCl@ss:
39021923
UNSPSC Code:
12161700
EC Number:
221-618-9
MDL number:
Beilstein/REAXYS Number:
6120352
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥98% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

pH

6.0-7.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.052, λ: 280 nm Amax: 0.039

SMILES string

N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6.2H3N/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);2*1H3/t1-,2-;;/m1../s1

InChI key

NGPGDYLVALNKEG-OLXYHTOASA-N

Application

Ammonium tartrate dibasic can be used to study biochemicals and reagents, biological buffers, chelators, protein structural analysis, proteomics and X-ray crystallography. Ammonium tartrate dibasic has been used in a study to research the crystal optimization and preliminary diffraction data analysis of the Smad1 MH1 domain bound to a palindromic SBE DNA element.


Still not finding the right product?

Explore all of our products under Ammonium tartrate dibasic


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library