Skip to Content
MilliporeSigma

09935

Ammonium pyrrolidinedithiocarbamate

≥98.0% (NT), powder with lumps, puriss. p.a.

Synonym(s):

1-Pyrrolidinecarbodithioic acid ammonium salt, APDC, Ammonium pyrrolidinecarbodithioate, PDC, PDTC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H9NS2 · NH3
CAS Number:
Molecular Weight:
164.29
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
225-834-4
MDL number:
Beilstein/REAXYS Number:
3730472
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ammonium pyrrolidinedithiocarbamate, purum p.a., ≥98.0% (NT)

grade

purum p.a.

Quality Segment

assay

≥98.0% (NT)

form

(powder or powder with lumps)

mp

153-155 °C (lit.)

cation traces

Bi: ≤10 mg/kg, Ca: ≤10 mg/kg, Cd: ≤10 mg/kg, Co: ≤10 mg/kg, Cr: ≤10 mg/kg, Cu: ≤10 mg/kg, Fe: ≤10 mg/kg, K: ≤50 mg/kg, Mg: ≤10 mg/kg, Mn: ≤10 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤10 mg/kg, Zn: ≤10 mg/kg

SMILES string

N.SC(=S)N1CCCC1

InChI

1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3

InChI key

MDDIUTVUBYEEEM-UHFFFAOYSA-N

Gene Information

human ... RELA(5970)

Application

for the precipitation of As, Bi, Cd, Co, Cu, Fe, Mn, Ni, Pb, Sb, Sn, V, Zn from acidic solutions

Other Notes

Reagent used for the extraction and pre-concentration of heavy metals with an organic solvent from aqueous solutions for atomic absorption spectroscopy, AAS. MIBK (methyl isobutyl ketone) is the solvent of choice; The extract can be directly injected into the flame


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library