Skip to Content
MilliporeSigma
All Photos(2)

Documents

08896

Supelco

2,4,6-Trichlorophenol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
776729
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

1 mmHg ( 76.5 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

bp

246 °C (lit.)

mp

64-66 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Héctor M Poggi-Varaldo et al.
Journal of environmental management, 113, 527-537 (2012-05-01)
The purpose of our research was to evaluate the effect of eliminating supplementation of sucrose to the reactor influent on the performance of a lab scale partially-aerated methanogenic fluidized bed bioreactor (PAM-FBBR). Two operational stages were distinguished: in the first
Meiqin Hu et al.
Environmental science & technology, 46(16), 9005-9011 (2012-07-26)
Molecular iodine has been studied, for the first time, as a sensitizer for the degradation of 2,4,6-trichlorophenol (TCP) in aqueous solution under visible light (λ ≥ 450 nm). TCP was degraded in the presence of commercial I(2), but the reaction
Stefan Franzen et al.
The journal of physical chemistry. B, 116(5), 1666-1676 (2011-12-14)
Kinetic and structural studies have shown that peroxidases are capable of the oxidation of 2,4,6-trichlorophenol (2,4,6-TCP) to 2,6-dichloro-1,4-benzoquinone (2,6-DCQ). Further reactions of 2,6-DCQ in the presence of H(2)O(2) and OH(-) yield 2,6-dichloro-3-hydroxy-1,4-benzoquinone (2,6-DCQOH). The reactions of 2,6-DCQ have been monitored
Xiao-Chi Feng et al.
Bioresource technology, 143, 642-646 (2013-07-17)
The present study investigated the synergistic effects of a novel combined uncoupler of TCS and TCP on excess activated sludge reduction during a 60-day operation using a sequence batch reactor (SBR). Response surface methodology (RSM) was employed to obtain the
Supreet S Bahga et al.
Electrophoresis, 33(6), 1048-1059 (2012-04-25)
We present a novel method of creating concentration cascade of leading electrolyte (LE) in isotachophoresis (ITP) by using bidirectional ITP. ITP establishes ion-concentration shock waves between high-mobility LE and low-mobility trailing electrolyte (TE) ions. In bidirectional ITP, we set up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service