Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08471

Supelco

Dichlorodimethylsilane solution

~5% in toluene, for GC derivatization, LiChropur

Synonym(s):

Sylon CT, Dimethyldichlorosilane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6Cl2Si
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

concentration

~5% in toluene

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.491-1.495

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For deactivating glassware.
Treat the glass tubing that you use for columns with Sylon-CT solution (5% dimethyldichlorosilane in toluene). It deactivates the tubing for use up to 350 °C to 400 °C, far exceeding the capabilities of other deactivating agents used at room temperature. Sylon CT also deactivates transfer lines and glass or glass-lined injection port liners. Instructions included.

Application

Learn more in the Product Information
Sylon CT was used as derivatizing agent, during an experiment to develop sensitive GC method for the identification and determination of long-chain primary alkyl amines.† It was also used for treating Erlenmeyer flasks to prevent mycelium from sticking to the glassware during development of techniques in the identification and partial characterization the chaxamycin biosynthesis gene cluster of S. leeuwenhoekii.†

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Sylon is a trademark of Sigma-Aldrich Co. LLC
Sylon CT is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service