Skip to Content
MilliporeSigma

07969

O-(2-Aminoethyl)polyethylene glycol 3,000

Mp 3,000

Synonym(s):

Aminopolyethylene glycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2(CH2CH2O)nH
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

Mp 3,000

Quality Segment

reaction suitability

reagent type: cross-linking reagent

Ω-end

hydroxyl

α-end

amine

SMILES string

[H]OCCN

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

General description

O-(2-Aminoethyl)polyethylene glycol is a heterobifunctional polyethylene glycol (hydroxyl-PEG-amine) useful in conjugation and crosslinking reactions.

Application

O-(2-Aminoethyl)polyethylene glycol has been used to synthesize PEG-poly(D,L-lactide-co-glycolide) (PEG-PLGA) conjugate for use in the synthesis of PLGA-based nanoparticles.

Legal Information

dPEG is a registered trademark of Quanta BioDesign


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library