Skip to Content
MilliporeSigma

07697

Fmoc-D-Glu(OtBu)-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-D-glutamic acid 5-tert-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H27NO6
CAS Number:
Molecular Weight:
425.47
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Assay:
≥98.0% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D +4.5±1, c = 1 in acetic acid: water (4:1)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

mp

86-96 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)CC[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C24H27NO6/c1-24(2,3)31-21(26)13-12-20(22(27)28)25-23(29)30-14-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,25,29)(H,27,28)/t20-/m1/s1

InChI key

OTKXCALUHMPIGM-HXUWFJFHSA-N



Still not finding the right product?

Explore all of our products under Fmoc-D-Glu(OtBu)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
289647-5G04061826284346
289647-25G04061826284339
07697-5G-F04061832641034
07697-1G-F04061826221266