Skip to Content
MilliporeSigma
All Photos(1)

Documents

07607

Supelco

Hydrochloric acid

0.5 M in methanol, for GC derivatization, LiChropur

Synonym(s):

Methanolic HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.05

product name

Hydrogen chloride solution, 0.5 M in methanol, for GC derivatization, LiChropur

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

concentration

0.45-0.55 M in methanol
0.5 M in methanol

technique(s)

gas chromatography (GC): suitable

density

0.801-0.812 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methanolic HCl was used in fatty acid analysis during a study to identify Δ4 fatty acid desaturated from Thraustochytrium sp. involved in the biosynthesis of docosahexanoic acid.

Learn more in the Product Information

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service