Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07411

Millipore

Xylose Disks

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Proteus spp.
Salmonella spp.
bacteria

Application

Xylose Disks are used to differentiate bacteria on the basis of carbohydrate fermentation abilities.

Principle

Xylose disks are used based on the principle that microorganisms can only ferment certain carbohydrates. The ability to cleave various carbohydrates depends on the type of enzymes they possess. This can be detected either by gas production (CO2) in liquid media or change in color of the pH indicator because of the acid production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service