Skip to Content
MilliporeSigma

06753

Isododecane

mixture of isomers, technical, ≥80% (GC)

Synonym(s):

2-Methylundecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H26
CAS Number:
Molecular Weight:
170.33
PubChem Substance ID:
UNSPSC Code:
12352100
NACRES:
NA.22
EC Number:
250-816-8
Assay:
≥80% (GC)
Technique(s):
GC/GC: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

technical

assay

≥80% (GC)

form

liquid

technique(s)

GC/GC: suitable

SMILES string

CC(CC(C)(C)C)CC(C)(C)C

InChI

1S/C12H26/c1-4-5-6-7-8-9-10-11-12(2)3/h12H,4-11H2,1-3H3

InChI key

GTJOHISYCKPIMT-UHFFFAOYSA-N



pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library