Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05156

Supelco

Albendazole-2-aminosulfone

analytical standard

Synonym(s):

2-Amino-5-propylsulfonylbenzimidazole, 2-Aminoalbendazole sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O2S
CAS Number:
Molecular Weight:
239.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

SMILES string

CCCS(=O)(C1=CC=C(N=C(N)N2)C2=C1)=O

InChI

1S/C10H13N3O2S/c1-2-5-16(14,15)7-3-4-8-9(6-7)13-10(11)12-8/h3-4,6H,2,5H2,1H3,(H3,11,12,13)

InChI key

WTPBIYSMFKUQKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole-2-aminosulfone is a metabolite of albendazole which is a broad-spectrum antihelmintic agent.

Application

Albendazole-2-aminosulfone may be used as a marker in the determination of albendazole residues in bovine milk using liquid-liquid extraction followed by reversed-phase high-performance liquid chromatography coupled with a fluorometric detector (RP-HPLC). It may be used as a reference standard in the determination of albendazole-2-aminosulfone in tissue samples using ultra performance liquid chromatography (UPLC).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of albendazole-2-aminosulfone in bovine milk using high-performance liquid chromatography with fluorometric detection
Chu SP, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 620(1), 129-135 (1993)
Simultaneous determination of albendazole and its metabolites in fish muscle tissue by stable isotope dilution ultra-performance liquid chromatography tandem mass spectrometry
Zhang X, et al.
Analytical and Bioanalytical Chemistry, 401(2), 727-734 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service