Skip to Content
MilliporeSigma

04683

Abberior® STAR 635P, streptavidin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12171501
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Abberior GmbH

fluorescence

λex 638 nm; λem 651 nm±10 nm in H2O

suitability

corresponds for SDS-PAGE

detection method

fluorometric

storage temp.

2-8°C

General description

Abberior® STAR 635P was the first member of an entirely new class of phosphorylated fluorescent dyes, which excels in its photophysical parameters. The product, Abberior® STAR 635P, streptavidin is a conjugate of the Abberior® STAR 635P type of dyes. It can be excited with lasers at wavelengths between 600 - 700 nm. For STED, a depletion wavelength around 750 - 800 nm is recommended. Abberior STAR 635P is the ideal partner for abberior STAR 580 or abberior STAR ORANGE to obtain optimal 2 color STED results. It can substitute ATTO 647N, Alexa Fluor 647.

Application

Designed and tested for fluorescent super-resolution microscopy

Features and Benefits

  • High brightness.
  • High photostability.
  • High water solubility enabling background-free imaging.
  • Suitable for fluorescence labeling of biotin-conjugated molecules.

Analysis Note

Unconjugated dye ≤5% of total fluorescence

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
abberior is a registered trademark of Abberior GmbH


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library