Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04646

Sigma-Aldrich

Hamamelitannin

≥98.0% (HPLC)

Synonym(s):

2-C-(Hydroxymethyl)-D-ribofuranose 2′,5-digallate, Hamamelofuranose 2′,5-digallate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O14
CAS Number:
Molecular Weight:
484.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

powder or crystals

application(s)

food and beverages
metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1O[C@H](COC(=O)c2cc(O)c(O)c(O)c2)[C@@H](O)[C@]1(O)COC(=O)c3cc(O)c(O)c(O)c3

InChI

1S/C20H20O14/c21-9-1-7(2-10(22)14(9)25)17(28)32-5-13-16(27)20(31,19(30)34-13)6-33-18(29)8-3-11(23)15(26)12(24)4-8/h1-4,13,16,19,21-27,30-31H,5-6H2/t13-,16-,19?,20-/m1/s1

InChI key

FEPAFOYQTIEEIS-IZUGRSKYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hamamelitanninis a tannin derived from the bark and leaves of Hamamelis virginiana(witch hazel).

Application

Hamamelitannin can be used to study chromatography, aromatics, esters, hamamelis, heterocyclics, natural compounds, phenols, phytopharma standards, polyhydroxy compounds, and tannins. It also significantly reduces biofilm metabolic activity of the following bacteria: Staphylococcus epidermidis, Staphylococcus aureus, Acinetobacter baumannii, and Candida albicans strains. Hamamelitannin displays specific cytotoxic activity against colon cancer cells. It has been used in a study to determine that quorum-sensing inhibitors increase the success of antibiotic treatment by increasing the susceptibility of bacterial biofilms and/or by increasing host survival following infection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Cobrado et al.
The Journal of antimicrobial chemotherapy, 67(5), 1159-1162 (2012-02-10)
The colonization of indwelling medical devices and subsequent biofilm formation represents a global challenge since it promotes the persistence of infection and contributes to antimicrobial resistance. The aim of this study was to determine the antimicrobial activity of cerium, chitosan
K Saeki et al.
Planta medica, 65(3), 227-229 (1999-05-08)
Tetragalloylglucose (TgG) and digalloylhamamelose (DgH) were found to inhibit adhesion to and invasion through Matrigel of mouse Lewis lung carcinoma LL2-Lu3 cells, which are highly metastatic. TgG inhibited matrix metalloproteinases (MMPs) from the tumor cells like (-)-epigallocatechin gallate, whereas DgH
Gilles Brackman et al.
Antimicrobial agents and chemotherapy, 55(6), 2655-2661 (2011-03-23)
Although the exact role of quorum sensing (QS) in various stages of biofilm formation, maturation, and dispersal and in biofilm resistance is not entirely clear, the use of QS inhibitors (QSI) has been proposed as a potential antibiofilm strategy. We
Solomon Habtemariam
Toxicon : official journal of the International Society on Toxinology, 40(1), 83-88 (2001-10-17)
The tumour necrosis factor-alpha (TNF) inhibitory activity of hamamelitannin from Hamamelis virginiana was investigated by assessing the TNF-mediated EAhy926 endothelial cell death and adhesiveness to monocytes. Treatment of the cells by TNF (25 ng/ml) and actinomycin D (0.1ng/ml) resulted in
H Masaki et al.
Journal of dermatological science, 10(1), 25-34 (1995-07-01)
The protective activities of hamamelitannin (2',5-di-O-galloyl-hamamelose) in Hamamelis virginiana L. and its related compound, gallic acid, on damaged murine skin fibroblasts induced by UVB irradiation were investigated. In order to exclude the UV absorbing effect of the compounds, the protection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service