Skip to Content
MilliporeSigma

04487

Bisphenol E

analytical standard

Synonym(s):

4,4′-Ethylidenebisphenol, 1,1-Bis(4-hydroxyphenyl)ethane, Bisphenol E

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH(C6H4OH)2
CAS Number:
Molecular Weight:
214.26
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

123-127 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C14H14O2/c1-10(11-2-6-13(15)7-3-11)12-4-8-14(16)9-5-12/h2-10,15-16H,1H3

InChI key

HCNHNBLSNVSJTJ-UHFFFAOYSA-N

Application

Bisphenol E may be used as a reference standard in the determination of bisphenol E in beverages using on-line solid-phase extraction with fast liquid chromatography coupled with tandem mass spectrometry (on-line SPE fast LC-MS/MS).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Analysis of bisphenols in soft drinks by on-line solid phase extraction fast liquid chromatography-tandem mass spectrometry.
Gallart-Ayala, et al.
Analytica Chimica Acta, 683(2), 227-233 (2011)



Global Trade Item Number

SKUGTIN
819016025004022536448664
819016010004022536448657
116025-50G04061838704023
819016999904022536448671
04487-100MG04061838632258