Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03694

Sigma-Aldrich

L-β-Homoserine

≥98.0% (TLC)

Synonym(s):

(R)-3-Amino-4-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
Beilstein:
7770197
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

application(s)

peptide synthesis

SMILES string

N[C@@H](CO)CC(O)=O

InChI

1S/C4H9NO3/c5-3(2-6)1-4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m1/s1

InChI key

BUZICZZQJDLXJN-GSVOUGTGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service