Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03630

Sigma-Aldrich

Ethylenediaminetetraacetic acid calcium disodium salt hydrate

≥97.0% (calc. based on dry substance, KT)

Synonym(s):

Calcium disodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OOCCH2)2NCH2CH2N(CH2COONa)2 · aq
CAS Number:
Molecular Weight:
374.27 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97.0% (calc. based on dry substance, KT)
form:
powder

Quality Level

Assay

≥97.0% (calc. based on dry substance, KT)

form

powder

functional group

amine

SMILES string

O.[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Ca.2Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;+2;2*+1;/p-4

InChI key

JHECKPXUCKQCSH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid calcium disodium salt hydrate (EDTA-CaNa2) is a chelating agent used in various fields including water treatment and pharmaceutical applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temmei Ito et al.
Biomedical materials (Bristol, England), 10(1), 015025-015025 (2015-03-03)
In order to obtain a hydroxyapatite (HAp)-poly(ethylene glycol) (PEG) composite, tetra amine-terminated PEG was crosslinked using disuccinimidyl tartrate to obtain a PEG hydrogel. Using two kinds of chelators with different stability constants for Ca ion (N-(2-hydroxyethyl) ethylenediamine-N,N',N'-triacetic acid (EDTA-OH, 8.14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service