Skip to Content
MilliporeSigma

02743

Sulfaethoxypyridazine

analytical standard

Synonym(s):

4-Amino-N-(6-ethoxy-3-pyridazinyl)benzenesulfonamide, N-1-(6-Ethoxy-3-pyridazinyl)sulfanilamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14N4O3S
CAS Number:
Molecular Weight:
294.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
213-514-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC), ≥97.0%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

CCOc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1

InChI

1S/C12H14N4O3S/c1-2-19-12-8-7-11(14-15-12)16-20(17,18)10-5-3-9(13)4-6-10/h3-8H,2,13H2,1H3,(H,14,16)

InChI key

FFJIWWBSBCOKLS-UHFFFAOYSA-N

General description

Chemical structure: sulfonamide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library