Skip to Content
MilliporeSigma
All Photos(2)

Documents

02351

Supelco

Poly(methacrylic acid sodium salt)

analytical standard, for GPC, 1,270

Synonym(s):

Poly(sodium methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~1100
Mp ~1250
Mw ~1270

Mw/Mn

~1.15

storage temp.

2-8°C

InChI

1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1

InChI key

SONHXMAHPHADTF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Poly(methacrylic acid sodium salt) is used as an analytical standard in the development of nebulized amphotericin B-polymethacrylic acid nanoparticles for use as pre-exposure prophylaxis agents to deliver anti-microbial drugs to the lung and inhibition of fungal infections caused by Aspergillus species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nebulised amphotericin B-polymethacrylic acid nanoparticle prophylaxis prevents invasive aspergillosis
Shirkhani K, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 11(5), 1217-1226 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service