Skip to Content

02150

Bis(1-butylpentyl) adipate

Selectophore, ≥98.0%

Synonym(s):

Adipic acid di-5-nonyl ester, BBPA, Di(5-nonyl) adipate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H46O4
CAS Number:
Molecular Weight:
398.62
UNSPSC Code:
26111700
NACRES:
NB.61
PubChem Substance ID:
EC Number:
278-791-9
Beilstein/REAXYS Number:
6572484
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

for ion-selective electrodes

Quality Segment

product line

Selectophore

assay

≥98.0%, ≥98.0% (GC)

form

liquid

density

0.910 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CCCC)OC(=O)CCCCC(=O)OC(CCCC)CCCC

InChI

1S/C24H46O4/c1-5-9-15-21(16-10-6-2)27-23(25)19-13-14-20-24(26)28-22(17-11-7-3)18-12-8-4/h21-22H,5-20H2,1-4H3

InChI key

VXQKWLPFTKRXCQ-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer employed in polymeric membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library