Skip to Content
MilliporeSigma

01993

Salmonella Chromogen Agar Set

suitable for microbiology, NutriSelect® Basic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171606
EC Number:
200-338-0
NACRES:
NA.85
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Basic

pH

7.3±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages, microbiology

storage temp.

2-8°C

suitability

nonselective and differential for Escherichia coli, nonselective and differential for coliforms, selective and differential for Escherichia coli, selective and differential for Klebsiella spp., selective and differential for Proteus spp., selective and differential for Salmonella spp., selective and differential for Shigella spp., selective and differential for coliforms, selective and differential for enterobacteriaceae

Application

A differential diagnostic agar for the detection of Salmonella in food and clinical material.

Preparation Note

Dissolve 29 g Salmonella Chromogen Agar (Cat. No. 84369) and 1 vial Salmonella Chromogen Agar Supplement (Cat. No. 38589) in 1 litre distilled water. Boil and shake for 35-40 minutes to dissolve the medium completely. Do not autoclave or overheat. Cool the medium as fast as possible to 45-50°C and shake gently for a further 30 minutes or less. Pour into cool petri dishes.

Other Notes

Medium Base (4x29g; Cat. No. 84369):
Peptone 5g/l
Yeast extract 2g/l
Meat extract 1g/l
Sodium chloride 5g/l
Sodium deoxycholate 1g/l
Agar 15g/l

4 vials of supplement (Cat. No. 38589; per vial, sufficient for 1000 ml medium):
chromogenic mix 0.13g
Propylenglycol 10.0g
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library