Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-1012

Sigma-Aldrich

Methyl Red solution

pH 4.2-6.2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Beilstein:
1843037
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

form

liquid

availability

available only in Japan

pH

4.2-6.2

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)/b17-16+

InChI key

CEQFOVLGLXCDCX-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oana Alexandru et al.
Journal of neuro-oncology, 102(1), 9-18 (2010-07-17)
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of
Chan-Ju Wang et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 66(Pt 1), 2-7 (2010-01-09)
Azoreductase 1 from Pseudomonas aeruginosa strain PAO1 (paAzoR1) catalyses the activation of the prodrug balsalazide and reduces the azo dye methyl red using reduced nicotinamide adenine dinucleotide cofactor as an electron donor. To investigate the mechanism of the enzyme, a
Sofia Babanova et al.
Journal of bioscience and bioengineering, 112(4), 379-387 (2011-07-26)
Soluble artificial mediators are often applied to enhance the electron transfer from living cells to an anode in microbial fuel cells. Recently, we have demonstrated that the Candida melibiosica 2491 yeast strain possesses electrogenic properties and can be used as
X D Zhang et al.
Journal of hazardous materials, 170(2-3), 883-887 (2009-06-09)
An aqueous C.I. Acid Red 2 solution was decolorized by electrolysis using iron as anode. The decolorization mechanism was investigated through experimental observations on the electrochemical behavior of C.I. Acid Red 2 on Pt rotating disk electrode, UV-visible spectra of
Danmeng Shuai et al.
Environmental science & technology, 44(5), 1773-1779 (2010-02-11)
Azo dyes are widespread pollutants and potential cocontaminants for nitrate; we evaluated their effect on catalytic reduction of a suite of oxyanions, diatrizoate, and N-nitrosodimethylamine (NDMA). The azo dye methyl orange significantly enhanced (less than or equal to a factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service