Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-5439

Sigma-Aldrich

Turbidity standard solution

turbidity 100 NTU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

form

liquid

availability

available only in Japan

concentration

0.1 mg/mL kaolin

turbidity

100 NTU

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service