Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-0980

Sigma-Aldrich

Silicon dioxide

JIS special grade

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

form

solid

availability

available only in Japan

refractive index

n20/D 1.544 (lit.)

mp

1610 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soania Mathur et al.
Journal of Parkinson's disease, 5(2), 263-268 (2015-02-28)
Despite an urgent need for new medications, clinical trials in Parkinson's have a relatively low rate of success. Although many reasons have been proposed for this, the opinions of patients and scientists, the two principal stakeholders, have not been widely
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which
Manuela Piazzi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1383-1394 (2015-01-01)
The phosphoinositide-dependent signal transduction pathway has been implicated in the control of a variety of biologic processes, such as the regulation of cellular metabolism and homeostasis, cell proliferation and differentiation, and apoptosis. One of the key players in the regulation
Luis Alberto Luévano-Martínez et al.
Biochimica et biophysica acta, 1847(6-7), 587-598 (2015-04-07)
Mitochondria play a key role in adaptation during stressing situations. Cardiolipin, the main anionic phospholipid in mitochondrial membranes, is expected to be a determinant in this adaptive mechanism since it modulates the activity of most membrane proteins. Here, we used
Mukesh K Jaiswal et al.
Journal of reproductive immunology, 112, 38-45 (2015-08-01)
The a2 isoform of vacuolar-ATPase (ATP6V0A2, referred to as a2V) is required for normal spermatogenesis and maturation of sperm. Treatment of male mice with anti-a2V disturbs the testicular cytokine/chemokine balance and leads to severe deficiencies of spermatogenesis. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service