Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5860

Sigma-Aldrich

L-Proline

SAJ special grade, ≥99.0%

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
grade:
SAJ special grade
Assay:
≥99.0%

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

228 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Couturier et al.
The Journal of biological chemistry, 288(20), 14624-14635 (2013-04-06)
The microbial deconstruction of the plant cell wall is a key biological process that is of increasing importance with the development of a sustainable biofuel industry. The glycoside hydrolase families GH5 (PaMan5A) and GH26 (PaMan26A) endo-β-1,4-mannanases from the coprophilic ascomycete
Salvatore Petta et al.
Hepatology (Baltimore, Md.), 59(5), 1692-1705 (2014-04-03)
We assessed the cost-effectiveness of sofosbuvir (SOF)-based triple therapy (TT) compared with boceprevir (BOC)- and telaprevir (TVR)-based TT in untreated genotype 1 (G1) chronic hepatitis C (CHC) patients discriminated according to IL28B genotype, severity of liver fibrosis, and G1 subtype.
Michael P Manns et al.
Gastroenterology, 147(2), 366-376 (2014-04-15)
MK-5172 is an inhibitor of the hepatitis C virus (HCV) nonstructural protein 3/4A protease; MK-5172 is taken once daily and has a higher potency and barrier to resistance than licensed protease inhibitors. We investigated the efficacy and tolerability of MK-5172
David Saadoun et al.
Annals of the rheumatic diseases, 73(5), 831-837 (2013-04-23)
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of
R A Wevers et al.
Clinical chemistry, 40(7 Pt 1), 1245-1250 (1994-07-01)
Although spin-echo techniques are often used to obtain 1H-NMR spectra of serum or plasma samples, they do not provide reliable quantitative analyses of metabolites. We present a standardized procedure, optimized for sensitivity, for using single-pulse 1H-NMR spectroscopy to analyze deproteinized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service