Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4520

Sigma-Aldrich

Potassium dichromate solution

1/60 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: oxidant

availability

available only in Japan

concentration

1/10 N
1/60 M

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter A Siver et al.
American journal of botany, 102(6), 921-941 (2015-06-24)
Heterokont algae of the class Synurophyceae, characterized by distinctive siliceous scales that cover the surface of the cell, are ecologically important in inland waters, yet their evolutionary history remains enigmatic. We explore phylogenetic relationships within this group of algae relative
Lubna Shakir et al.
Environmental toxicology and pharmacology, 34(2), 180-191 (2012-04-24)
The problem of water pollution acquires greater relevance in the context of a developing agrarian economy like Pakistan. Even though, the leather industry is a leading economic sector in Pakistan, there is an increasing environmental concern regarding tanneries because they
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement
Shafaqat Ali et al.
Environmental toxicology and chemistry, 32(10), 2234-2239 (2013-06-19)
A hydroponic experiment was carried out to examine the effect of hydrogen sulfide (H2 S) in alleviating chromium (Cr) stress in barley. A 2-factorial design with 6 replications was selected, including 3 levels of NaHS (0 μM, 100 μM, and
B Duarte et al.
Ecotoxicology and environmental safety, 83, 1-7 (2012-07-13)
The in situ reduction of Cr (VI) to its less toxic form Cr (III) may be a useful detoxification mechanism for phytoremediation. Using a hydroponics mesocosmos approach, we evaluated the ability of Halimione portulacoides to reduce and uptake Cr (VI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service