Skip to Content
MilliporeSigma

24-3745

Poly(vinyl alcohol)

SAJ first grade, degree of polymerization 500

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
UNSPSC Code:
12162002
MDL number:
Grade:
SAJ first grade
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

SAJ first grade

form

solid

mol wt

degree of polymerization 500

availability

available only in Japan

dilution

(for analytical testing)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Poly(vinyl alcohol)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library