Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-0570

Sigma-Aldrich

Paraffin liquid

SAJ first grade

Synonym(s):

Mineral oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
grade:
SAJ first grade
technique(s):
immunohistochemistry: suitable

grade

SAJ first grade

form

liquid

availability

available only in Japan

technique(s)

immunohistochemistry: suitable

refractive index

n20/D 1.467 (lit.)

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

460.4 °F

Flash Point(C)

238 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Endo et al.
Plant physiology, 147(4), 1984-1993 (2008-06-14)
The regulation of abscisic acid (ABA) biosynthesis is essential for plant responses to drought stress. In this study, we examined the tissue-specific localization of ABA biosynthetic enzymes in turgid and dehydrated Arabidopsis (Arabidopsis thaliana) plants using specific antibodies against 9-cis-epoxycarotenoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service