Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11-0398

Sigma-Aldrich

Fluorescamine

≥98.0%

Synonym(s):

4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10O4
CAS Number:
Molecular Weight:
278.26
Beilstein:
921143
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
Assay:
≥98.0%

Assay

≥98.0%

availability

available only in Japan

mp

153-157 °C (lit.)

SMILES string

O=C1OC2(OC=C(C2=O)c3ccccc3)c4ccccc14

InChI

1S/C17H10O4/c18-15-13(11-6-2-1-3-7-11)10-20-17(15)14-9-5-4-8-12(14)16(19)21-17/h1-10H

InChI key

ZFKJVJIDPQDDFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides to form stable, highly fluorescent compounds. Low background due to hydrolysis. Useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes. Effectively blocks newly generated amino termini in protein sequence analyses.

Suitability

for HPLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire E Stanley et al.
Chimia, 66(3), 88-98 (2012-05-02)
This account highlights some of our recent activities focused on developing microfluidic technologies for application in high-throughput and high-information content chemical and biological analysis. Specifically, we discuss the use of continuous and segmented flow microfluidics for artificial membrane formation, the
Khin Yin Win et al.
The Analyst, 137(10), 2328-2332 (2012-03-13)
Here we report a fast and effective method to visualize interactive proteins across intact mammalian cells via on-site formation of fluorescence using instant reaction of non-fluorescent fluorescamine with primary amines on proteins. Without interference by fluorescence background, this fluorogenic labelling
Andrew D Aubrey et al.
Astrobiology, 8(3), 583-595 (2008-08-06)
The Urey organic and oxidant detector consists of a suite of instruments designed to search for several classes of organic molecules in the martian regolith and ascertain whether these compounds were produced by biotic or abiotic processes using chirality measurements.
Yusuke Suzuki et al.
Journal of agricultural and food chemistry, 56(22), 10811-10816 (2008-11-06)
Photoinduced decarboxylation via homolytic cleavage of the ester linkage generating two benzyl radicals being recoupled is known to be a major photolytic pathway of the insecticide fenvalerate in aqueous or organic solvents. A highly sensitive and selective fluorescence spectroscopic method
Min Jia et al.
Analytical chemistry, 81(19), 8033-8040 (2009-10-01)
Fluorescamine derivatized 3-amino-2,2,5,5,-tetramethyl-1-pyrrolidinyloxy (I) is shown to undergo an irreversible reaction with peroxyl radicals and other radical oxidants to generate a more highly fluorescent diamagnetic product (II) and thus can be used as a highly sensitive and versatile probe to

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service